Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BG_PPAP
Entrez Gene ID
834215
Gene Name
beta-1,3-glucanase
Gene Aliases
3-GLUCANASE_PUTATIVE, 3-glucanase_putative, ARABIDOPSIS THALIANA BETA-1, AT5G42100, ATBG_PAP, ATBG_PPAP, MJC20.21, MJC20_21, beta-1
Chromosome Location
Chr.5: 16829113 - 16831618 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123575.4 NP_199025.1 1 - 2 1656 69
GenBank mRNA AY054690.1 - 1 - 2 1234 69
AY084866.1 - 1 - 2 1230 69
BT008863.1 - 1 - 2 1206 69

View Full Product Details