Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUL4
Entrez Gene ID
834663
Gene Name
cullin4
Gene Aliases
AT5G46210, ATCUL4, MDE13.3, MDE13_3, cullin4
Chromosome Location
Chr.5: 18731371 - 18736815 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123990.3 NP_568658.1 15 - 16 2347 111
GenBank mRNA AY084267.1 - 15 - 16 2206 111

View Full Product Details