Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OPR1
Entrez Gene ID
844001
Gene Name
12-oxophytodienoate reductase 1
Gene Aliases
12-OXOPHYTODIENOATE REDUCTASE 1, 12-oxophytodienoate reductase 1, ARABIDOPSIS 12-OXOPHYTODIENOATE REDUCTASE 1, AT1G76680, ATOPR1, F28O16.5, F28O16_5
Chromosome Location
Chr.1: 28776626 - 28778837 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106318.4 NP_177794.1 1 - 2 570 128
NM_202428.2 NP_974157.1 1 - 2 335 128
GenBank mRNA AY074874.1 - 1 - 2 284 128
BT020365.1 - 1 - 2 214 128

View Full Product Details