The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_115822.4 | NP_191519.1 | 3 - 4 | 378 | 133 |
GenBank mRNA | AK119016.1 | - | 3 - 4 | 350 | 133 |
AY086002.1 | - | 3 - 4 | 370 | 133 | |
BT008531.1 | - | 2 - 3 | 238 | 133 |