Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PEG3
Entrez Gene ID
444864
Gene Name
paternally expressed 3
Gene Aliases
-
Chromosome Location
Chr.18: 64099192 - 64122748 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002887.3 NP_001002887.3 1 - 2 143 65
GenBank mRNA AY427787.1 - - 141 65

View Full Product Details