Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TADA3
Entrez Gene ID
510184
Gene Name
transcriptional adaptor 3
Gene Aliases
TADA3L
Chromosome Location
Chr.22: 16923587 - 16933845 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024511.1 NP_001019682.1 7 - 8 1008 107
GenBank mRNA BT020627.1 - 7 - 8 1008 107

View Full Product Details