Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP6V0D1
Entrez Gene ID
282148
Gene Name
ATPase H+ transporting V0 subunit d1
Gene Aliases
ATP6D, ATP6DV
Chromosome Location
Chr.18: 35046554 - 35085620 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174505.3 NP_776930.1 7 - 8 944 62
GenBank mRNA BC102305.1 - 7 - 8 971 62
J04204.1 - 7 - 8 945 62

View Full Product Details