Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD28
Entrez Gene ID
281050
Gene Name
CD28 molecule
Gene Aliases
-
Chromosome Location
Chr.2: 91906337 - 91944271 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181004.1 NP_851347.1 3 - 4 533 69
GenBank mRNA X93304.1 - - 533 69

View Full Product Details