Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTUB2
Entrez Gene ID
504880
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 2
Gene Aliases
-
Chromosome Location
Chr.21: 58636677 - 58659061 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015517.1 NP_001015517.1 2 - 3 138 86
GenBank mRNA BT020786.1 - 2 - 3 138 86

View Full Product Details