Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM9B
Entrez Gene ID
515665
Gene Name
TMEM9 domain family member B
Gene Aliases
-
Chromosome Location
Chr.15: 43625137 - 43643809 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034507.2 NP_001029679.1 2 - 3 297 138
GenBank mRNA BC102506.1 - 2 - 3 298 138

View Full Product Details