Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SDF4
Entrez Gene ID
528783
Gene Name
stromal cell derived factor 4
Gene Aliases
-
Chromosome Location
Chr.16: 51270260 - 51282976 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035375.1 NP_001030452.1 5 - 6 856 71
GenBank mRNA BC103026.1 - 5 - 6 856 71

View Full Product Details