Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VOPP1
Entrez Gene ID
767861
Gene Name
VOPP1 WW domain binding protein
Gene Aliases
ECOP
Chromosome Location
Chr.22: 514680 - 694820 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076972.1 NP_001070440.1 4 - 5 416 143
GenBank mRNA BC149078.1 - 4 - 5 442 143
BT021736.1 - 4 - 5 416 143

View Full Product Details