Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A7
Entrez Gene ID
767975
Gene Name
membrane-spanning 4-domains, subfamily A, member 7
Gene Aliases
-
Chromosome Location
Chr.15: 83184411 - 83210498 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077040.1 NP_001070508.1 5 - 6 685 137
GenBank mRNA BT021572.1 - 5 - 6 685 137

View Full Product Details