Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMAB
Entrez Gene ID
617636
Gene Name
metabolism of cobalamin associated B
Gene Aliases
ATR
Chromosome Location
Chr.17: 63628778 - 63639339 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079632.1 NP_001073100.1 2 - 3 233 81
GenBank mRNA BC111194.1 - 2 - 3 184 81
BT021748.1 - 2 - 3 233 81

View Full Product Details