Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CALR
Entrez Gene ID
281036
Gene Name
calreticulin
Gene Aliases
-
Chromosome Location
Chr.7: 12618114 - 12621944 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174000.2 NP_776425.1 2 - 3 266 58
GenBank mRNA AB067687.1 - 2 - 3 266 58
BC140582.1 - 2 - 3 259 58

View Full Product Details