Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CES2
Entrez Gene ID
505928
Gene Name
carboxylesterase 2
Gene Aliases
-
Chromosome Location
Chr.18: 34638138 - 34650199 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034260.2 NP_001029432.1 10 - 11 1515 101
GenBank mRNA BC102288.1 - 10 - 11 1516 101

View Full Product Details