Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG5
Entrez Gene ID
532686
Gene Name
autophagy related 5
Gene Aliases
-
Chromosome Location
Chr.9: 43654728 - 43807560 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034579.2 NP_001029751.2 1 - 2 49 120
GenBank mRNA BC102770.1 - 1 - 2 49 120

View Full Product Details