Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPI
Entrez Gene ID
513586
Gene Name
mannose phosphate isomerase
Gene Aliases
-
Chromosome Location
Chr.21: 33797779 - 33805138 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035284.1 NP_001030361.1 1 - 2 41 67
GenBank mRNA BC102844.1 - 1 - 2 41 67

View Full Product Details