Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPI
Entrez Gene ID
513586
Gene Name
mannose phosphate isomerase
Gene Aliases
-
Chromosome Location
Chr.21: 33797779 - 33805138 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035284.1 NP_001030361.1 3 - 4 375 60
GenBank mRNA BC102844.1 - 3 - 4 375 60

View Full Product Details