Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL26
Entrez Gene ID
514636
Gene Name
methyltransferase like 26
Gene Aliases
C25H16orf13
Chromosome Location
Chr.25: 544097 - 545876 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038106.2 NP_001033195.1 5 - 6 748 122
GenBank mRNA BC109870.1 - 5 - 6 750 122

View Full Product Details