Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMRTC2
Entrez Gene ID
539134
Gene Name
DMRT like family C2
Gene Aliases
Dmrt7
Chromosome Location
Chr.18: 51367680 - 51372973 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038182.2 NP_001033271.1 7 - 8 886 89
GenBank mRNA BC109620.1 - 7 - 8 887 89

View Full Product Details