Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM53
Entrez Gene ID
540738
Gene Name
transmembrane protein 53
Gene Aliases
-
Chromosome Location
Chr.3: 101355002 - 101371875 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038572.1 NP_001033661.1 2 - 3 224 92
GenBank mRNA BC109854.1 - 2 - 3 224 92

View Full Product Details