Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4A3
Entrez Gene ID
515145
Gene Name
eukaryotic translation initiation factor 4A3
Gene Aliases
-
Chromosome Location
Chr.19: 52468433 - 52480178 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046188.2 NP_001039653.1 4 - 5 420 70
GenBank mRNA BC111184.1 - 4 - 5 421 70

View Full Product Details