Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAT1L
Entrez Gene ID
618809
Gene Name
vesicle amine transport 1 like
Gene Aliases
-
Chromosome Location
Chr.18: 5004884 - 5169822 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046624.2 NP_001040089.1 6 - 7 1010 101
GenBank mRNA BC114121.1 - 6 - 7 1011 101

View Full Product Details