Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YJU2
Entrez Gene ID
514934
Gene Name
YJU2 splicing factor homolog
Gene Aliases
CCDC94
Chromosome Location
Chr.7: 19796423 - 19812076 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075691.1 NP_001069159.1 4 - 5 469 138
GenBank mRNA BC118290.1 - 4 - 5 469 138

View Full Product Details