Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS20
Entrez Gene ID
614441
Gene Name
regulator of G protein signaling 20
Gene Aliases
RSG20
Chromosome Location
Chr.14: 21924527 - 21955493 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076327.1 NP_001069795.1 1 - 2 237 75
GenBank mRNA BC116015.1 - 1 - 2 237 75

View Full Product Details