Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRKRIP1
Entrez Gene ID
780786
Gene Name
PRKR interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.25: 34630465 - 34659008 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079641.2 NP_001073109.1 4 - 5 436 102
GenBank mRNA BC112521.1 - 4 - 5 437 102

View Full Product Details