Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HID1
Entrez Gene ID
540436
Gene Name
HID1 domain containing
Gene Aliases
C19H17orf28
Chromosome Location
Chr.19: 56455021 - 56475765 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076924.1 NP_001070392.1 3 - 4 447 72
GenBank mRNA BC123459.1 - 3 - 4 447 72

View Full Product Details