Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMC10
Entrez Gene ID
507692
Gene Name
ER membrane protein complex subunit 10
Gene Aliases
C18H19orf63, INM02
Chromosome Location
Chr.18: 56613682 - 56620090 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080239.1 NP_001073708.1 2 - 3 234 145
GenBank mRNA BC123856.1 - 2 - 3 204 145
BC126720.1 - 2 - 3 234 145

View Full Product Details