Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM38A
Entrez Gene ID
532775
Gene Name
transmembrane protein 38A
Gene Aliases
-
Chromosome Location
Chr.7: 6356470 - 6382966 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083463.1 NP_001076932.1 3 - 4 479 75
GenBank mRNA BC123825.1 - 3 - 4 479 75

View Full Product Details