Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF2IRD2
Entrez Gene ID
539745
Gene Name
GTF2I repeat domain containing 2
Gene Aliases
-
Chromosome Location
Chr.25: 32703586 - 32750409 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083745.1 NP_001077214.1 5 - 6 720 85
GenBank mRNA BC134477.1 - 5 - 6 720 85

View Full Product Details