Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YIPF7
Entrez Gene ID
522186
Gene Name
Yip1 domain family member 7
Gene Aliases
-
Chromosome Location
Chr.6: 63238857 - 63290362 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103258.1 NP_001096728.1 4 - 5 526 93
GenBank mRNA BC140593.1 - 4 - 5 526 93

View Full Product Details