Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H11orf34
Entrez Gene ID
505518
Gene Name
chromosome 15 open reading frame, human C11orf34
Gene Aliases
PLET1
Chromosome Location
Chr.15: 22642076 - 22654185 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113538.1 NP_001107010.1 2 - 3 424 114
GenBank mRNA BC140681.1 - 2 - 3 424 114
EE252825.1 - 2 - 3 425 114

View Full Product Details