Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UTP11
Entrez Gene ID
537675
Gene Name
UTP11 small subunit processome component
Gene Aliases
-
Chromosome Location
Chr.3: 107941135 - 107957939 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098092.1 NP_001091561.1 5 - 6 432 103
GenBank mRNA BC140486.1 - 5 - 6 432 103

View Full Product Details