Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TWNK
Entrez Gene ID
538467
Gene Name
twinkle mtDNA helicase
Gene Aliases
C26H10orf2, twinkle
Chromosome Location
Chr.26: 21824850 - 21830238 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098463.1 NP_001091933.1 2 - 3 2048 119
GenBank mRNA BC140487.1 - 2 - 3 2048 119

View Full Product Details