Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPPA3
Entrez Gene ID
616433
Gene Name
developmental pluripotency associated 3
Gene Aliases
-
Chromosome Location
Chr.5: 101333531 - 101341323 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111108.2 NP_001104578.1 2 - 3 283 61
NM_001111109.2 NP_001104579.1 1 - 2 532 61
GenBank mRNA EF446904.1 - 1 - 2 532 61
EF446905.1 - 2 - 3 283 61
EU143548.1 - 1 - 2 33 61

View Full Product Details