Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELANE
Entrez Gene ID
100126050
Gene Name
elastase, neutrophil expressed
Gene Aliases
ELA2
Chromosome Location
Chr.7: 43411323 - 43413728 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105653.1 NP_001099123.1 4 - 5 625 96
GenBank mRNA BC149420.1 - 4 - 5 625 96

View Full Product Details