Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTHD1
Entrez Gene ID
538470
Gene Name
ENTH domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 111158863 - 111256820 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205542.1 NP_001192471.1 3 - 4 711 81

View Full Product Details