Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL15
Entrez Gene ID
540246
Gene Name
kelch like family member 15
Gene Aliases
-
Chromosome Location
Chr.X: 119240097 - 119277860 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205799.1 NP_001192728.1 1 - 2 711 102

View Full Product Details