Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADARB2
Entrez Gene ID
524061
Gene Name
adenosine deaminase RNA specific B2 (inactive)
Gene Aliases
-
Chromosome Location
Chr.13: 46039693 - 46272683 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192588.3 NP_001179517.1 2 - 3 185 66

View Full Product Details