Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD80
Entrez Gene ID
407131
Gene Name
CD80 molecule
Gene Aliases
-
Chromosome Location
Chr.1: 64296500 - 64326059 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206439.1 NP_001193368.1 1 - 2 103 72
GenBank mRNA DY081230.1 - 3 - 4 218 72
Y09950.1 - - 98 72

View Full Product Details