Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALDH5A1
Entrez Gene ID
532724
Gene Name
aldehyde dehydrogenase 5 family member A1
Gene Aliases
-
Chromosome Location
Chr.23: 33181628 - 33209663 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192735.1 NP_001179664.1 4 - 5 705 98

View Full Product Details