Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGI2
Entrez Gene ID
541046
Gene Name
leucine rich repeat LGI family member 2
Gene Aliases
-
Chromosome Location
Chr.6: 44589538 - 44617975 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192816.1 NP_001179745.1 5 - 6 484 87

View Full Product Details