Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF12
Entrez Gene ID
100140477
Gene Name
Kruppel like factor 12
Gene Aliases
-
Chromosome Location
Chr.12: 48526365 - 48802837 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191270.2 NP_001178199.2 3 - 4 672 99

View Full Product Details