Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KSR2
Entrez Gene ID
617325
Gene Name
kinase suppressor of ras 2
Gene Aliases
-
Chromosome Location
Chr.17: 57231429 - 57668251 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206862.1 NP_001193791.1 4 - 5 985 63

View Full Product Details