Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRIT3
Entrez Gene ID
511618
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 3
Gene Aliases
-
Chromosome Location
Chr.6: 15552132 - 15566741 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191514.2 NP_001178443.1 1 - 2 445 87

View Full Product Details