Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECT2L
Entrez Gene ID
100141063
Gene Name
epithelial cell transforming 2 like
Gene Aliases
-
Chromosome Location
Chr.9: 76563428 - 76639348 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191539.1 NP_001178468.1 8 - 9 1197 86

View Full Product Details