Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLR8
Entrez Gene ID
532262
Gene Name
toll like receptor 8
Gene Aliases
-
Chromosome Location
Chr.X: 130728235 - 130745827 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033937.1 NP_001029109.1 2 - 2 2918 100
GenBank mRNA AY642125.2 - 2 - 2 2918 100
EF583902.1 - 1 - 1 2945 100
GU936182.1 - 1 - 1 2945 100
GU936183.1 - 1 - 1 2945 100

View Full Product Details