Product Details

SNP ID
rs76912569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187370872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACGTACTGGGAGCACTGGACATC[C/G]CAGGGTCAGGTGCGTATGAGGCTCT
Phenotype
MIM: 609350
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RTP4 PubMed Links

Gene Details

Gene
RTP4
Gene Name
receptor transporter protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022147.2 302 Silent Mutation TCC,TCG S80S NP_071430.2

View Full Product Details