Product Details

SNP ID
rs78547906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:188688848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGACTTTCCCACTAGATTGTGAG[C/T]TCCTGGAGGGCAGGCACTTTCGTTC
Phenotype
MIM: 600700 MIM: 612154
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPP PubMed Links

Gene Details

Gene
LPP
Gene Name
LIM domain containing preferred translocation partner in lipoma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167671.2 Intron NP_001161143.1
NM_001167672.2 Intron NP_001161144.1
NM_005578.4 Intron NP_005569.1
XM_005247446.4 Intron XP_005247503.1
XM_005247450.4 Intron XP_005247507.1
XM_005247451.4 Intron XP_005247508.1
XM_005247453.2 Intron XP_005247510.1
XM_011512820.2 Intron XP_011511122.1
XM_011512823.2 Intron XP_011511125.1
XM_011512827.2 Intron XP_011511129.1
XM_011512828.2 Intron XP_011511130.1
XM_011512831.2 Intron XP_011511133.1
XM_011512833.2 Intron XP_011511135.1
XM_011512834.2 Intron XP_011511136.1
XM_011512836.2 Intron XP_011511138.1
XM_017006377.1 Intron XP_016861866.1
XM_017006378.1 Intron XP_016861867.1
XM_017006379.1 Intron XP_016861868.1
XM_017006380.1 Intron XP_016861869.1
XM_017006381.1 Intron XP_016861870.1
XM_017006382.1 Intron XP_016861871.1
Gene
MIR28
Gene Name
microRNA 28
There are no transcripts associated with this gene.

View Full Product Details